Balls
Earth model beach ball, Ø: 9" inflated, 16" deflated, —
Catalog # 470003-348
Supplier: Skywill Far East Industries
Supplier #: 2048500
Jump to:
Product Details & Documents
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.
Specifications
- Désignation:Earth model beach ball, Ø: 9" inflated, 16" deflated
- Diameter:—
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



