Radiation Lab Activity
Radiation Lab Activity
Radiation Lab Activity, Materials for Eight Lab Groups, 8
Catalog # 470002-500
Radiation Lab Activity
Catalog # 470002-500
Supplier:  
Radiation Lab Activity
Radiation Lab Activity
Radiation Lab Activity, Materials for Eight Lab Groups, 8
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
KIT RADIATION CAN WITH ACTIVITY GUIDE Catalog #470304-034
Product 470304-034 is a functional alternative to 470002-500 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Product Details & Documents
Students investigate the absorption and radiation of energy on different surfaces.
  • Activity Includes Light Source
  • Two Experiments in One Activity
  • Students Research Absorption and Radiation of Energy
They then turn on the light at 10 cm distance and observe how temperatures respond differently in the black can and silver can.
In the reverse experiment, the rates of cooling can be determined after students turn off the light.
Ordering information: Each kit includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, and two other thermometers (-20° to 50°C). The single kit also includes one 120VAC clamp lamp with a reflector and 100 watt light bulb; the class kit includes four lamps.
Specifications
  • Désignation:
    Radiation Lab Activity, Materials for Eight Lab Groups
  • No. of Lab Groups:
    8
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamp lamp with reflector
    Clamp lamp with reflector
    470004-224
    Log in for availability
    Each - $45.95