Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dissection Placemats | Avantor

Dissection Placemats

Fetal Pig Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm
Catalog # 470002-398
Supplier:   Ward's Science
Fournisseur #:  420.5040.1
Dissection Placemats
Catalog # 470002-398
Supplier:  Ward's Science
Dissection Placemats
Dissection Placemats

Fetal Pig Dissection Placemat, Polyvinyl, 40 cm, 24.7 cm

Price
21,35 $
Log in pour la disponibilité
Each
Product Details & Documents

Perfect Instructional Companion for Wet Lab Areas

  • Resistant to spills of any kind

Illustrations and descriptive text are printed right on the durable polyvinyl mat, so students can have specimen classification and information on the internal and external anatomy right on the lab bench with them while they are dissecting. Can be easily cleaned with a sponge or a paper towel, so students don’t have to worry about damaging a text book or other paper copies.

Spécifications
  • Largeur:
    24.7 cm
  • Désignation:
    Fetal Pig Dissection Placemat
  • Longueur:
    40 cm
  • Finish:
    Polyvinyl