Dissolved Oxygen
LaMotte® Classroom Dissolved Oxygen Test Kit, Refill Kit
  470021-308
 :   LaMotte
 :  3976A-H
Dissolved Oxygen
  470021-308
 :  LaMotte
Dissolved Oxygen
Dissolved Oxygen
LaMotte® Classroom Dissolved Oxygen Test Kit, Refill Kit
$28.60
Each
Enough materials for five groups working simultaneously.
  • Contains enough materials for a total of 25 tests
  • The kit will measure 0, 4, and 8 ppm
Provide your students with hands-on reinforcement of key concepts in water quality and water treatment with this test kit. Includes a teacher’s manual with lesson plans, background information, and reproducible student procedure and data sheets.
  • Désignation:
    LaMotte® Classroom Dissolved Oxygen Test Kit, Refill Kit