Direct Reading Titrators
Direct Reading Titrator
Catalog # 470021-210
Supplier: LaMotte
Supplier #: 0382
Jump to:
Product Details & Documents
Safe alternative to Glass Titrators
- Various Volumes
- Easy to Read
Chemically-resistant plastic eliminates the inconvenience of glass breakage, while the precision tip increases sensitivity. It is available in three different ranges.
Specifications
- Désignation:Direct Reading Titrator