LaMotte® Salinity Test Kit | Avantor
LaMotte® Salinity Test Kit
Refill Kit for LaMotte™ Salinity Water Test Kit
Catalog # 470021-206
Supplier:   LaMotte
Fournisseur #:  R-7459-01
LaMotte® Salinity Test Kit
Catalog # 470021-206
Supplier:  LaMotte
CAS Number:  
LaMotte® Salinity Test Kit
LaMotte® Salinity Test Kit
Refill Kit for LaMotte™ Salinity Water Test Kit
CAS Number:  
Price
$103.25
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit test for salinity using the Direct Reading Titrator method.
  • Aligned with GLOBE protocols
Spécifications
Spécifications
  • Désignation:
    Refill Kit for LaMotte™ Salinity Water Test Kit