Nitrate-Nitrogen | Avantor
Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Catalog # 470021-202
Fournisseur #:  3354-01
Nitrate-Nitrogen
Catalog # 470021-202
Supplier:  
Nitrate-Nitrogen
Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
DET OF NITRITE NITRATE KIT Catalog #470219-152
Le produit 470219-152 est similaire à 470021-202 mais le fabricant et les spécifications peuvent varier.
NITRATE TEST KIT, MODEL NI-11 Catalog #CA11014-426
Le produit CA11014-426 est similaire à 470021-202 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
  • Range: 0 to 15 ppm
  • Material for 50 tests
  • Compact, waterproof carrying case contains kit
  • Instructions included
  • Pre-calibrated test tubes
  • Aligned with GLOBE protocols
Spécifications
  • Désignation:
    LaMotte® Water Test Kit, Nitrate-Nitrogen