Polypropylene Beaker Sets
250 / 500 / 1000 ml, Set of 3, 25 / 50 / 100 ml
Catalog # 470020-820
Supplier: Dynalon
Fournisseur #: 522135-0003
Jump to:
Product Details & Documents
Unbreakable Polypropylene
- Autoclavable
- Chemical-resistant
These beaker sets are autoclavable up to 145°C and chemical resistant. All feature pour spouts and metric graduation scales.
Spécifications
- Intervalle de graduation:25 / 50 / 100 ml
- Capacité:250 / 500 / 1000 ml
- Désignation:Set of 3