Medium Grade, 15 μm
Filter Paper, Medium Grade, 125 mm diameter
Catalog # 470019-886
Supplier: United Scientific Supplies
: FPR125
Jump to:
Product Details & Documents
The most widely used qualitative filter paper for routine applications.
- Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
: Packages of 100.
Spécifications
- Désignation:Filter Paper, Medium Grade, 125 mm diameter



