Lab Label Roll
Lab label roll, dispenser, ¾", 100"
Catalog # 470019-780
Fournisseur #: 130575
Jump to:
Discontinued
Product Details & Documents
Better than gummed labels, these pressure-sensitive labels adhere securely, yet can be easily removed.
- Autoclavable to 260 °C
The labels are on a continuous roll.
Spécifications
- Désignation:Lab label roll, dispenser
- Longueur:100"
- Width:¾"