Specific Gravity Hydrometer
Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions
Catalog # 470019-566
Supplier: G & W Instruments
Fournisseur #: A-1101
Jump to:
Product Details & Documents
Use these hydrometers with all processes where specific gravity or density of liquids is an important control.
The design of this hydrometer ensures easy cleaning and a low likelihood of breakage. The clear markings allow accurate readings. Accuracy: ±0.001. Size: 12"L.
Spécifications
- Désignation:Specific Gravity Hydrometer, 1.00–1.20, 0.002 divisions