Right Angle Clamp Holders
Right Angle Clamp Holder, for 1/2" dia. Rods
Catalog # 470019-546
Supplier: Humboldt Manufacturing
Fournisseur #: H-8300
Jump to:
Product Details & Documents
The perpendicular V-shaped jaws of these clamp holders are desiged to support rods and clamp extensions at a 90° angle.
Holders feature thumbscrews at a 180° angle.
Spécifications
- Désignation:Right Angle Clamp Holder, for 1/2" dia. Rods