Mayo Scissors
Mayo Scissors, 172 mm (6.75"), Stainless steel, Sharp/Blunt
Catalog # 470018-948
Supplier: Dunrite Instruments
Fournisseur #: 140992
Jump to:
Product Details & Documents
Safe yet sharp.
- Single blunt side prevents tissue damage
- Available in 2 lengths
- Stainless construction resists rust
Designed for use in the famous clinic, these straight-blade scissors allow for precise control. They are offered in two lengths. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Mayo Scissors
- Longueur:172 mm (6.75")
- Matière:Stainless steel
- Blade Type:Sharp/Blunt