Crystal Models
Graphite
Catalog # 470018-082
Supplier: United Scientific Supplies
Fournisseur #: CMSGRP
Jump to:
Product Details & Documents
Gorgeous Visual of Crystals
- Introduce Crystal Lattices to your Students
- Available in Three Types
These beautiful large crystal models enable you to introduce crystal lattices and unit cells to your classroom. Not only are they durable and provide for a great visual, they are also mounted permanently onto a clear acrylic base to minimize the possibility of lost parts.
Spécifications
- Désignation:Graphite