Backpack Lab Soil Quality Educational Test Kit
Backpack Lab Soil Quality Educational Test Kit
Backpack lab soil quality educational test kit, 0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)
Catalog # 470017-358
Supplier #:  HI3896BP
Backpack Lab Soil Quality Educational Test Kit
Catalog # 470017-358
Supplier:  
CAS Number:  
Backpack Lab Soil Quality Educational Test Kit
Backpack Lab Soil Quality Educational Test Kit
Backpack lab soil quality educational test kit, 0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Backpack Lab designed for educators and environmental science students, containing lessons, activities, and test kits that relate to important parameters evaluated in soil quality. 
  • Test kits for the determination of common parameters in soil quality testing: nitrogen, phosphorus, potassium and pH.
Backpack Lab is designed with all the necessary components in one place, reducing the chance of misplacing an item. Ideal for transporting, it's easy to take this durable backpack to the field for on-site measurements. This portable kit contains well-constructed lessons and activities, and will allow the teacher to get the most out of their classroom time. The Combo pH/EC/TDS Tester is also included for measurement of pH, conductivity (EC), and total dissolved solids (TDS), as well as the T-Shaped Thermometer for accurate temperature measurements.
Specifications
Specifications
  • Désignation:
    Backpack lab soil quality educational test kit
  • Range:
    0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)
  • Cat. No.:
    470017-358
  • Accuracy:
    ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
  • Calibration:
    Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
  • Resolution:
    0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)