Document Analysis Lab Activity
Analysis of Documents Kit Refill, 6
Catalog # 470015-368
Supplier:   Kemtec
 :  10-506R
Document Analysis Lab Activity
Catalog # 470015-368
Supplier:  Kemtec
Document Analysis Lab Activity
Document Analysis Lab Activity
Analysis of Documents Kit Refill, 6
Price
$339.85
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.
Spécifications
  • Désignation:
    Analysis of Documents Kit Refill
  • No. of Lab Groups:
    6