Gas Generating Bottle Set
Gas Generating Bottle Set, Capacity: 250 mL
Catalog # 470007-660
Supplier: GSC International
Supplier #: 9001
Jump to:
Product Details & Documents
When used with a pneumatic trough, our gas generating bottle set is ideal for generating small quantities of hydrogen and other gases.
- Hydrogen gas production requires 3M HCl and mossy zinc
The set consists of a bottle, 2-hole rubber stopper, reagent addition (thistle) tube, and gas delivery tube.
Ordering information: Pneumatic trough, ¹/₄" Int.Ø rubber tubing, and gas collection bottles are sold separately.
Specifications
- Désignation:Gas Generating Bottle Set, Capacity: 250 mL