Snap Circuits Educational Electronics Training Programs | Avantor
Snap Circuits Educational Electronics Training Programs
SC300R Educational Electronics Training Program
Catalog # 470007-232
Supplier:   Elenco Electronics
Fournisseur #:  SC300R
Snap Circuits Educational Electronics Training Programs
Catalog # 470007-232
Supplier:  Elenco Electronics
Snap Circuits Educational Electronics Training Programs
Snap Circuits Educational Electronics Training Programs
SC300R Educational Electronics Training Program
Price
$336.90
Log in pour la disponibilité
Each
Product Details & Documents
The award-winning Snap Circuit Series uses building blocks with snaps to build electrical and electronic circuit projects.
  • Experiment Manual Included
  • Five Hundred Experiments Included
  • Storage Box Included
  • Experiment Topics Include Basic Components and Circuits, Capacitors, and More
  • Series Uses Building Blocks with Snaps to Build Electrical Projects
Each block has a function, and the blocks are color coded and numbered so they're easy to identify.
The blocks feature 500 experiments including basic components and circuits, motors and electricity, resistance, capacitors, transistors, oscillators and electronic sound, integrated circuits, electromagnetism, and radio. Other experiments cover meters, transformers, and FM radio; diodes and applications; and electronic switches.
Ordering information: This kit includes an experiment manual, student guide, and parts organizer/storage box. It requires two AA batteries (not included).
Spécifications
  • Désignation:
    SC300R Educational Electronics Training Program
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.