Assorted Rubber Stoppers | Avantor
Assorted Rubber Stoppers
00 to 8
Catalog # 470005-802
Assorted Rubber Stoppers
Catalog # 470005-802
Supplier:  
Assorted Rubber Stoppers
Assorted Rubber Stoppers
00 to 8
Discontinued
Product Details & Documents
Solid, 1-hole and 2-hole stoppers in assorted sizes.
  • Superior grade of rubber
  • Soft, resilent, and durable
  • Low sulfur content
  • Size numbers molded onto the top
  • All stoppers are 25mm long
  • Aligned with GLOBE protocols
Designed to stay pliable over long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.
Caution: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.
Spécifications
  • Taille:
    00 to 8