Assorted Rubber Stoppers
00 to 8
Catalog # 470005-802
Jump to:
Discontinued
Product Details & Documents
Solid, 1-hole and 2-hole stoppers in assorted sizes.
- Superior grade of rubber
- Soft, resilent, and durable
- Low sulfur content
- Size numbers molded onto the top
- All stoppers are 25mm long
- Aligned with GLOBE protocols
Designed to stay pliable over long period of time, these stoppers can withstand temperatures from –25 to 70°C (–13 to 158°F). Ideal for use with nonaggressive or concentrated solutions.
Caution: Item contains latex, which is known to cause allergic reactions in some people. Use with caution.
Spécifications
- Taille:00 to 8
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.