Multi Purpose Fire Extinguishers
Multi-purpose fire extinguisher, 2.5 lbs., 1A10B:C
  470005-488
 :  21032136K
Multi Purpose Fire Extinguishers
  470005-488
Multi Purpose Fire Extinguishers
Multi-purpose fire extinguisher, 2.5 lbs., 1A10B:C
$148.30
Each
For use on Fire Types (Classes) A, B, and C.
  • Pressure gauge provides at-a-glance status
  • Easy-to-pull safety pin
  • Powder coated cylinder for corrosion protection
  • 6-year limited warranty
These ideal classroom fire extinguishers are effective against all three types of fire; wood and paper, liquids and grease, and electrical. Both extinguishers contain dry chemical powder, pressurized with carbon dioxide gas. The powder is discharged in a blanket-like stream at a range of more than four meters to smother the fire. Both extinguishers are supplied with wall-mounting hardware.
 : Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
  • Désignation:
    Multi-purpose fire extinguisher, 2.5 lbs.
  • UL Rating:
    1A10B:C