Replacement Bulbs
Replacement Bulb, with Bi-Pin Base, 12 V, 20 W
Catalog # 470005-028
Supplier: VWR
Fournisseur #: SE060098
Jump to:
Product Details & Documents
Replacement microscope light bulbs are available for a variety of microscopes. If unsure which bulb type is needed, please contact Ward's Science.
Spécifications
- Désignation:Replacement Bulb, with Bi-Pin Base
- Electrical:12 V, 20 W