Model Plus: Cells
Model Plus: Cells
Model Plus: The Plant Cell, 22 cm, 33 cm, 29 cm
  470004-828
Supplier #:  69074
Model Plus: Cells
  470004-828
Model Plus: Cells
Model Plus: Cells
Model Plus: The Plant Cell, 22 cm, 33 cm, 29 cm
$139.60
Each
Learn Cell Structure With Hands-On Foam Pieces
  • Easy To Manipulate Pieces
  • Guides Allow Students To Build And Learn At Their Own Pace
  • Designed For Grades 1-8
Simple for elementary level students yet comprehensive enough for middle school, these colorful teaching models Were designed specifically for hands-on use. Each model is made of resilient, non-toxic EVA foam to withstand years of active investigation. A thorough, detailed teacher’s guide includes inquiry-based lab activities, reproducible worksheets, complete background information, cross-curricular extension activities, assessment ideas and more. Topics covered include cell structure and function, osmosis, eukaryotes, bacteria and more.
  • Désignation:
    Model Plus: The Plant Cell
  • Hauteur:
    29 cm
  • Longueur:
    33 cm
  • Width:
    22 cm