Ohaus® Spring Scales
8003-PN - 0.05 lbs. (0.25 N) graduation intervals, 2.25 lbs. (10 N)
Catalog # 470004-634
Supplier:   OHAUS CORP.
 :  80000043
Ohaus® Spring Scales
Catalog # 470004-634
Supplier:  OHAUS CORP.
Ohaus® Spring Scales
Ohaus® Spring Scales
8003-PN - 0.05 lbs. (0.25 N) graduation intervals, 2.25 lbs. (10 N)
Price
$63.80
Log in pour la disponibilité
Each
Product Details & Documents
These spring scales are designed to stand up to heavy student use.
  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
Spécifications
  • Désignation:
    8003-PN - 0.05 lbs. (0.25 N) graduation intervals
  • Weighing Capacity:
    2.25 lbs. (10 N)