Document Analysis Lab Activity | Avantor
Document Analysis Lab Activity
Document Analysis Kit, 6
Catalog # 470002-510
Supplier:   Kemtec
Fournisseur #:  10-506
Document Analysis Lab Activity
Catalog # 470002-510
Supplier:  Kemtec
Document Analysis Lab Activity
Document Analysis Lab Activity
Document Analysis Kit, 6
Price
$528.60
Log in pour la disponibilité
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Determine a document’s authenticity.
  • Grades 6-12
  • Analysis of secret messages using chemistry
Your students will be fascinated as they perform both physical and chemical analysis of handwriting materials found at a crime scene. The procedures they will follow cover checking for erasures, abnormal positioning of a signature, use of different inks, and differences in handwriting or typing. It comes with enough materials for six setups and an instruction manual. A UV lamp, required, is available separately.
Spécifications
  • Désignation:
    Document Analysis Kit
  • No. of Lab Groups:
    6