Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Illustrated Dissection Guides | Avantor

Illustrated Dissection Guides

Mammalian Heart Guide, 10, 22 cm, 14 cm
Catalog # 470002-346
Supplier:   Ward's Science
Fournisseur #:  420.4174.1
Illustrated Dissection Guides
Catalog # 470002-346
Supplier:  Ward's Science
Illustrated Dissection Guides
Illustrated Dissection Guides

Mammalian Heart Guide, 10, 22 cm, 14 cm

Price
$24.75
Log in pour la disponibilité
Each
Product Details & Documents

These economical and concise guides contain the right amount of information for reference during dissection.

  • Basic Facts on Specimens
  • Convenient Size
  • Paperback
  • Color Illustrations
  • Step-by-Step Instructions

Each booklet provides basic facts on specimens, step-by-step dissection instructions, and clear illustrations to guide students as they dissect their own specimens.

The paperback guides are small, so students can have them whenever they're needed.

Spécifications
  • Désignation:
    Mammalian Heart Guide
  • Longueur:
    22 cm
  • No. of Pages:
    10
  • Width:
    14 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.