Nitrate-Nitrogen | Avantor
Nitrate-Nitrogen
Nitrate Refill Kit
Catalog # 470002-190
Fournisseur #:  R-3354
Nitrate-Nitrogen
Catalog # 470002-190
Supplier:  
Nitrate-Nitrogen
Nitrate-Nitrogen
Nitrate Refill Kit
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
  • Range: 0 to 15 ppm
  • Material for 50 tests
  • Compact, waterproof carrying case contains kit
  • Instructions included
  • Pre-calibrated test tubes
  • Aligned with GLOBE protocols
Spécifications
  • Désignation:
    Nitrate Refill Kit