Turbidity | Avantor
Turbidity
Turbidity Refill Kit
Catalog # 470002-188
Supplier:   LaMotte
Fournisseur #:  7520-H
Turbidity
Catalog # 470002-188
Supplier:  LaMotte
Turbidity
Turbidity
Turbidity Refill Kit
Price
$32.60
Log in pour la disponibilité
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: Five to 200 JTU
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
A compact, waterproof carrying case contains reagents, labware, and step-by-step instructions.
The tests have a range of 5 to 200 JTU.
Spécifications
  • Désignation:
    Turbidity Refill Kit