Held Secure™ Slide/Cassette Storage System, Electron Microscopy Sciences | Avantor
Held Secure™ Slide/Cassette Storage System, Electron Microscopy Sciences
Slide Index Cards
Catalog # 102773-600
Fournisseur #:  63280-54
Held Secure™ Slide/Cassette Storage System, Electron Microscopy Sciences
Catalog # 102773-600
Held Secure™ Slide/Cassette Storage System, Electron Microscopy Sciences
Slide Index Cards
Price
$61.39
Log in pour la disponibilité
Case of 500 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A durable corrugated filing box kit for the proper filing, identification, and storage of glass slides or cassettes.
  • Partitioned drawers for easy retrieval
  • Double tabbed in back for additional safety
  • Printing on box front designed for easy inventory identification
  • Internal corrugated construction designed for multiple stacking
  • Sold in unassembled cases of 10 boxes
  • 50 In/Out Cards come with each Slide/Cassette case for slide retrieval documentation
  • 25 Index Cards come with each Slide Kit for slide retrieval documentation
  • Manufactured in the USA
The complete set comes unassembled in packages of 10. Designed to hold up to 2,000 slides or 500+ cassettes per box. Additional cards and removable tray labels available for bulk purchase.
Spécifications
  • Désignation:
    Slide Index Cards