Held Secure™ Slide/Cassette Storage System, Electron Microscopy Sciences
Product Details & Documents
A durable corrugated filing box kit for the proper filing, identification, and storage of glass slides or cassettes.
- Partitioned drawers for easy retrieval
- Double tabbed in back for additional safety
- Printing on box front designed for easy inventory identification
- Internal corrugated construction designed for multiple stacking
- Sold in unassembled cases of 10 boxes
- 50 In/Out Cards come with each Slide/Cassette case for slide retrieval documentation
- 25 Index Cards come with each Slide Kit for slide retrieval documentation
- Manufactured in the USA
The complete set comes unassembled in packages of 10. Designed to hold up to 2,000 slides or 500+ cassettes per box. Additional cards and removable tray labels available for bulk purchase.
Spécifications
- Désignation:Hold Secure for Cassettes