Vacuum Pickup Kit
Vacuum pen kit, 3 star quality, includes PVS-CB-12, PVB-CB-12, CB-14, CB-38
89412-002
: Excelta
: PV-1-ESD-CNX
- Désignation:Vacuum pen kit, 3 star quality, includes PVS-CB-12, PVB-CB-12, CB-14, CB-38
- Cat. No.:89412-002
- Length:125 mm (5")
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- 63043-610Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.02", ID 0.005" cup sizeEach - 212,29 $
- Description
- Description
- Description
- 63043-616Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/2" (12.5 mm) dia. cup sizeEach - 13,99 $
- Description
- 63043-618Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/4" (6.25 mm) dia. cup sizeEach - 13,99 $
- Description
- 63043-622Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/32" (2.38 mm) dia. cup sizeEach - 14,84 $
- Description
- 63043-624Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/4" (18.75 mm) dia. cup sizeEach - 25,77 $
- Description
- 63043-626Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/8" (9.5 mm) dia. cup sizeEach - 13,86 $
- Description
- 63043-628Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 5/8" (15.75 mm) dia. cup sizeEach - 25,99 $
- Description
- 63043-644Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.02", ID 0.005" Cup SizeEach - 207,87 $
- Description
- 63043-646Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.03", ID 0.02" Cup SizeEach - 179,59 $
- Description
- 63043-648Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.05", ID 0.04" Cup SizeEach - 199,53 $
- Description
- 63043-650Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/2" (12.5 mm) dia. cup sizeEach - 13,45 $
- Description
- 63043-654Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/8" (3.25 mm) dia. cup sizeEach - 14,23 $
- Description
- 63043-656Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/32" (2.38 mm) dia. cup sizeEach - 13,71 $
- Description
- 63043-658Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/4" (18.75 mm) dia. cup sizeEach - 25,01 $
- Description
- 63043-660Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/8" (9.5 mm) dia. cup sizeEach - 13,32 $
- Description
- 63043-662Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 5/8" (15.75 mm) dia. cup sizeEach - 23,60 $
- Description
- Description
- 94024-430Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/16" (4.76 mm) dia. cup sizeEach - 12,26 $
- Description
- Description
- Description
- Description
- 63043-620Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/8" (3.25 mm) dia. cup sizeEach - 13,99 $
- Description
- Description
- Description
- Description
- Description