Vacuum Pickup Kit
Vacuum pen kit, 3 star quality, includes PVS-CB-12, PVB-CB-12, CB-14, CB-38
  89412-002
 :   Excelta
 :  PV-1-ESD-CNX
Vacuum Pickup Kit
  89412-002
 :  Excelta
Vacuum Pickup Kit
Vacuum Pickup Kit
Vacuum pen kit, 3 star quality, includes PVS-CB-12, PVB-CB-12, CB-14, CB-38
$251.94
Each
  • Désignation:
    Vacuum pen kit, 3 star quality, includes PVS-CB-12, PVB-CB-12, CB-14, CB-38
  • Cat. No.:
    89412-002
  • Length:
    125 mm (5")
Accessories
  •  
    Description
  • 63043-266
    Vacuum cup, black Buna-N, 3 star quality, 1/8" (3.25 mm) dia. cup size
    Each - 7,35 $
  •  
    Description
  • 63043-268
    Vacuum cup, black Buna-N, 3 star quality, 3/32" (2.38 mm) dia. cup size
    Each - 7,55 $
  •  
    Description
  • 63043-270
    Vacuum cup, black Buna-N, 3 star quality, 3/4" (18.75 mm) dia. cup size
    Each - 15,89 $
  •  
    Description
  • 63043-272
    Vacuum cup, black Buna-N, 3 star quality, 3/8" (9.5 mm) dia. cup size
    Each - 7,47 $
  •  
    Description
  • 63043-274
    Vacuum cup, black Buna-N, 3 star quality, 5/8" (15.75 mm) dia. cup size
    Each - 15,34 $
  •  
    Description
  • 63043-610
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.02", ID 0.005" cup size
    Each - 212,29 $
  •  
    Description
  • 63043-612
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.03", ID 0.02" cup size
    Each - 186,80 $
  •  
    Description
  • 63043-614
    Vacuum tip, acetal, 3 star quality, 30° bent; OD 0.05", ID 0.04" cup size
    Each - 179,85 $
  •  
    Description
  • 63043-616
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/2" (12.5 mm) dia. cup size
    Each - 13,99 $
  •  
    Description
  • 63043-618
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/4" (6.25 mm) dia. cup size
    Each - 13,99 $
  •  
    Description
  • 63043-622
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/32" (2.38 mm) dia. cup size
    Each - 14,84 $
  •  
    Description
  • 63043-624
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/4" (18.75 mm) dia. cup size
    Each - 25,77 $
  •  
    Description
  • 63043-626
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/8" (9.5 mm) dia. cup size
    Each - 13,86 $
  •  
    Description
  • 63043-628
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 5/8" (15.75 mm) dia. cup size
    Each - 25,99 $
  •  
    Description
  • 63043-644
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.02", ID 0.005" Cup Size
    Each - 207,87 $
  •  
    Description
  • 63043-646
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.03", ID 0.02" Cup Size
    Each - 179,59 $
  •  
    Description
  • 63043-648
    Vacuum probe/cup, acetal, 3 star quality, Straight; OD 0.05", ID 0.04" Cup Size
    Each - 199,53 $
  •  
    Description
  • 63043-650
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/2" (12.5 mm) dia. cup size
    Each - 13,45 $
  •  
    Description
  • 63043-654
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 1/8" (3.25 mm) dia. cup size
    Each - 14,23 $
  •  
    Description
  • 63043-656
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/32" (2.38 mm) dia. cup size
    Each - 13,71 $
  •  
    Description
  • 63043-658
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/4" (18.75 mm) dia. cup size
    Each - 25,01 $
  •  
    Description
  • 63043-660
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 3/8" (9.5 mm) dia. cup size
    Each - 13,32 $
  •  
    Description
  • 63043-662
    Vacuum probe/cup, black Buna-N, 3 star quality, Straight, 5/8" (15.75 mm) dia. cup size
    Each - 23,60 $
  •  
    Description
  • 94024-400
    Vacuum cup, black Buna-N, 3 star quality, 3/16" (4.76 mm) dia. cup size
    Each - 6,23 $
  •  
    Description
  • 94024-430
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 3/16" (4.76 mm) dia. cup size
    Each - 12,26 $
  •  
    Description
  • 94024-434
    Vacuum replacement bulb, 2 star quality
    Each - 38,18 $
  •  
    Description
  • 63043-262
    Vacuum cup, black Buna-N, 3 star quality, 1/2" (12.5 mm) dia. cup size
    Each - 7,35 $
  •  
    Description
  • 63043-264
    Vacuum cup, black Buna-N, 3 star quality, 1/4" (6.25 mm) dia. cup size
    Each - 7,10 $
  •  
    Description
  • 63043-620
    Vacuum probe/cup, black Buna-N, 3 star quality, 30° bent, 1/8" (3.25 mm) dia. cup size
    Each - 13,99 $
  •  
    Description
  • 75880-500
    Vacuum pen only, 3 star quality
    Each - 160,78 $
  •  
    Description
  • 75880-688
    Vacuum systems, hand piece, 4 star quality
    Each - 267,09 $
  •  
    Description
  • 75880-690
    Vacuum systems, air filter, 4 star quality
    Each - 33,05 $
  •  
    Description
  • 75880-692
    Vacuum systems, standard performance, 4 star quality
    Each - 1 213,61 $
  •  
    Description
  • 94024-438
    Vacuum pen only, 3 star quality
    Each - 206,06 $