CF® Dye Hydrazide, Biotium
CF®568 hydrazide, 562/583 nm, 1 mg
  89410-818
 :   Biotium
 :  92154
CF® Dye Hydrazide, Biotium
  89410-818
 :  Biotium
 :  
CF® Dye Hydrazide, Biotium
CF® Dye Hydrazide, Biotium
CF®568 hydrazide, 562/583 nm, 1 mg
 :  
$340.13
Each (1mg)
Fluorescent CF® Dye hydrazides are bright, extremely water-soluble, nontoxic molecules, useful as aldehyde-fixable cell tracers. They can also be used to label aldehyde or ketone groups of polysaccharides, carbohydrates or glycoproteins.
  • Fluorescently labeled low molecular weight, polar, non-toxic molecules; useful as aldehyde-fixable cell tracers
  • Membrane impermeant, can be easily loaded into cells by microinjection
  • Can be used to fluorescently label aldehyde or ketone groups of polysaccharides, carbohyrdates, or glycoproteins
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent imaging
Bright, extremely water-soluble and non-toxic CF® Dye hydrazides are excellent fluorescent tracers for neurons or gap junction studies. The cell impermeant tracer dyes can be introduced into cells by microinjection.
CF® Dye hydrazides can also be used to fluorescently label aldehyde or ketone groups of carbohydrates or glycoproteins molecules after peroxidation with periodate.
For carbohydrates labeling applications, we recommend using CF® Dye aminooxy forms, which are more reactive toward these groups than hydrazide forms. For fluorescence two-dimensional differential gel electrophoresis (2D-DIGE), we recommend CFDI hydrazides.
  • Désignation:
    CF®568 hydrazide
  • Taille:
    1 mg
  • Ex/Em:
    562/583 nm