CF® Dye Hydrazide, Biotium
CF® Dye Hydrazide, Biotium
CF®555 hydrazide, 555/565 nm, 1 mg
Catalog # 89410-816
Supplier:   Biotium
Supplier #:  92153
CF® Dye Hydrazide, Biotium
Catalog # 89410-816
Supplier:  Biotium
CAS Number:  
CF® Dye Hydrazide, Biotium
CF® Dye Hydrazide, Biotium
CF®555 hydrazide, 555/565 nm, 1 mg
CAS Number:  
Price
340,13 $
Log in pour la disponibilité
Each (1mg)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Fluorescent CF® Dye hydrazides are bright, extremely water-soluble, nontoxic molecules, useful as aldehyde-fixable cell tracers. They can also be used to label aldehyde or ketone groups of polysaccharides, carbohydrates or glycoproteins.
  • Fluorescently labeled low molecular weight, polar, non-toxic molecules; useful as aldehyde-fixable cell tracers
  • Membrane impermeant, can be easily loaded into cells by microinjection
  • Can be used to fluorescently label aldehyde or ketone groups of polysaccharides, carbohyrdates, or glycoproteins
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent imaging
Bright, extremely water-soluble and non-toxic CF® Dye hydrazides are excellent fluorescent tracers for neurons or gap junction studies. The cell impermeant tracer dyes can be introduced into cells by microinjection.
CF® Dye hydrazides can also be used to fluorescently label aldehyde or ketone groups of carbohydrates or glycoproteins molecules after peroxidation with periodate.
For carbohydrates labeling applications, we recommend using CF® Dye aminooxy forms, which are more reactive toward these groups than hydrazide forms. For fluorescence two-dimensional differential gel electrophoresis (2D-DIGE), we recommend CFDI hydrazides.
Spécifications
Spécifications
  • Désignation:
    CF®555 hydrazide
  • Taille:
    1 mg
  • Ex/Em:
    555/565 nm