Peroxide Test, MQuant®, MilliporeSigma
Test with Strips, Enzymatic reaction, 0,5-2-5-10-25 mg/l H₂O₂, 25 strips
Catalog # CA1.10011.0002
Supplier:   MilliporeSigma
 :  1.10011.0002
Peroxide Test, MQuant®, MilliporeSigma
Catalog # CA1.10011.0002
Supplier:  MilliporeSigma
CAS Number:  
Peroxide Test, MQuant®, MilliporeSigma
Test with Strips, Enzymatic reaction, 0,5-2-5-10-25 mg/l H₂O₂, 25 strips
CAS Number:  
Price
$96.47
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The peroxide test is ideal for testing residual concentrations of disinfectants on equipment or the product itself following cleaning and rinsing.
Specifications
Specifications
  • Méthode chimique:
    Enzymatic reaction
  • Désignation:
    Test with Strips
  • Range:
    0,5-2-5-10-25 mg/l H₂O₂
  • No. of Tests:
    25 strips