Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
2.5 kg, Plastic Bottle
Catalog # EM1.01090.2500
Supplier #:  1.01090.2500
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
Catalog # EM1.01090.2500
Supplier:  
CAS Number:  
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
2.5 kg, Plastic Bottle
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ALUMINIUM OXIDE 60 G NEUT ETLC Catalog #CA1.01090.2500
Product CA1.01090.2500 is a replacement for EM1.01090.2500 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Standardized sorbents ensure reliable results every time
EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
Specifications
Specifications
  • Type de conditionnement:
    Plastic Bottle
  • Taille:
    2.5 kg