BD Microtainer® Tubes, BD Diagnostics
BD Microtainer® tube, K2 EDTA, Lavender
Catalog # CABD365974L
Supplier:   BD
 :  365974
BD Microtainer® Tubes, BD Diagnostics
Catalog # CABD365974L
Supplier:  BD
BD Microtainer® Tubes, BD Diagnostics
BD Microtainer® Tubes, BD Diagnostics
BD Microtainer® tube, K2 EDTA, Lavender
Price
$97.80
Log in pour la disponibilité
Pack of 50 Case of 200
Product Details & Documents
BD Microtainer® tubes are designed for the collection, transport, and processing of skin puncture blood.
The “twist-assist” BD Microgard™ closure is ready-to-use and reduces blood splatter upon removal.
The integrated collection scoop, along with a wider diameter tube, allow for ease of collection and improved mixing. To simplify additive and sample identification, Microtainer® tubes are color-coded in a manner similar to evacuated tubes. Volume level lines, printed on tubes, ensure proper blood-to-anticoagulant ratio.
Spécifications
  • Désignation:
    BD Microtainer® tube
  • Additive:
    K2 EDTA
  • Cap Color:
    Lavender
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Tube Extender for use with VT365974
    Tube Extender for use with VT365974
    CABD365976
    Log in pour la disponibilité
    Pack of 50 - $66.70