pH Indicator en solution, large gamme pH 4-10
Dropping bottle, marked, 100 ml
Catalog # CA97026-686
Supplier: Hach
: 2329332
Jump to:
Product Details & Documents
For visual determination of pH, using test kits
Method: 10222
- ONU:1219
- MARC:3,II
Spécifications
Spécifications
- Type de conditionnement:Dropping bottle, marked
- Taille:100 ml