pH Buffer Solutions, Hach
±0.02, 25 °C, 500 mL, 7.00, Colorless
Catalog # CA97021-426
Supplier: Hach
: 1222249
Jump to:
Product Details & Documents
For pH meter standardization
Available colorless, or color-coded for easy identification.
Certificates
Search by Lot NumberSpecifications
Specifications
- Color:Colorless
- Taille:500 mL
- Reference Temperature:25 °C
- pH:7.00
- pH Accuracy:±0.02



