Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach | Avantor

SINGLET™ Single-Use pH Buffer, Combination Pack, Hach

±0.02, 7.00
Catalog # CA97021-424
Supplier:   Hach
Fournisseur #:  2770120
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
Catalog # CA97021-424
Supplier:  Hach
CAS Number:  
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach
SINGLET™ Single-Use pH Buffer, Combination Pack, Hach

±0.02, 7.00

CAS Number:  
Price
$146.00
Log in pour la disponibilité
Pack of 20
Product Details & Documents

Single-use standard solutions eliminate the need to prepare standardized buffer solutions for pH meter calibration

  • Ideal for on-site analysis
  • Easy to use
  • Fresh and reliable

You can have fresh pH buffer solutions without having to purchase and carry deionized or demineralized water. The convenient foil-pouch packaging maintains freshness, and keeps the solutions contaminant-free and ready to travel with you anywhere.

Ordering information: Single-Use pH Buffer Combination Packs are packaged with 20 pouches with 10 of each pH listed per pack.

Spécifications

Spécifications

  • pH:
    7.00
  • pH Accuracy:
    ±0.02
  • Cat. No.:
    CA97021-424
  • Concentration:
    7.00 pH

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.