VWR® Compound Trinocular Microscope
Product Details & Documents
VWR® Compound Laboratory Microscopes, equipped with Planachromatic Objectives, are ideal for laboratory, routine, and research work.
- Equipped with planachromatic objectives
- Ergonomic, anti-fatigue design
- Siedentopf-type head with a 30° inclination and 360° rotating capabilities
- One-year warranty on electrical parts
- Five-year warranty on mechanical parts and optics
The series offers an ergonomic, anti- fatigue design to enable long hours of comfortable observation.
This VWR® Compound Microscope features a Siedentopf type head with a 30° inclination and 360° rotating capabilities. Standard features include, widefield eyepieces WF10X / 20mm, reversed quintuple nosepiece, plan objectives PL 4X, 10X, 40X S, 100X S-Oil, coaxial coarse and fine focusing adjustment, built in low position coaxial mechanical stage, centerable and focusable 1.25 N.A Abbe condenser, iris diaphragm with filter holder, Koehler illumination halogen 12V / 20W with intensity control, main supply 110V UL approved.
Certifications: C-UL, UL Listed. CSA certified.
Ordering information: Microscope is supplied with all accessories listed above. The microscope is also supplied with a one-year warranty on electrical parts, and a five-year warranty on mechanical parts and optics.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:VWR® 2.5X SLR Projection Lens
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.