Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual micrometer calibration slide, 76 mm×25 mm, negative ima
Catalog # 102681-170
Supplier #:  68040-07
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Catalog # 102681-170
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual micrometer calibration slide, 76 mm×25 mm, negative ima
Price
$1,954.72
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS has introduced two new calibration slides that have the benefit of dual imperial/metric scales.
  • New Dual-Scale Calibration Slides
  • 2" Imperial (English) and 50mm Metric Scales on a Single Slide
  • Positive and Negative Versions
  • Unique Serial Number for Traceability
  • Available with Internationally Traceable Certificates of Calibration
The PS52P is for transmitted light applications and has a bright chrome positive image. The PS52N has a negative pattern, formed in low reflective chrome for incident light applications to give excellent contrast. Both are ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Standard and Crossed Patterns – S1, S2, S4, S5, S8, S11, S12, S16, S20, S21, S22, S48.
Spécifications
  • Désignation:
    Dual micrometer calibration slide, 76 mm×25 mm, negative ima