Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual micrometer calibration slide, 76 mm×25 mm, positive im
  102681-168
 :  68040-05
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
  102681-168
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual Micrometer Calibration Slide, Electron Microscopy Sciences
Dual micrometer calibration slide, 76 mm×25 mm, positive im
$1,598.12
Each
EMS has introduced two new calibration slides that have the benefit of dual imperial/metric scales.
  • New Dual-Scale Calibration Slides
  • 2" Imperial (English) and 50mm Metric Scales on a Single Slide
  • Positive and Negative Versions
  • Unique Serial Number for Traceability
  • Available with Internationally Traceable Certificates of Calibration
The PS52P is for transmitted light applications and has a bright chrome positive image. The PS52N has a negative pattern, formed in low reflective chrome for incident light applications to give excellent contrast. Both are ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Standard and Crossed Patterns – S1, S2, S4, S5, S8, S11, S12, S16, S20, S21, S22, S48.
  • Désignation:
    Dual micrometer calibration slide, 76 mm×25 mm, positive im