Air-O-Cell Cassette, Electron Microscopy Sciences | Avantor
Air-O-Cell Cassette, Electron Microscopy Sciences
Cassette
Catalog # 102681-058
Fournisseur #:  65210-50
Air-O-Cell Cassette, Electron Microscopy Sciences
Catalog # 102681-058
Air-O-Cell Cassette, Electron Microscopy Sciences
Air-O-Cell Cassette, Electron Microscopy Sciences
Cassette
Price
$831.19
Log in pour la disponibilité
Each Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is a unique air sampling cassette specially designed for quick collection of a wide range of airborne aerosols including mold spores, pollen, insect parts, skin cell fragments, fibers (e.g., asbestos, fiberglass, cellulose, clothing fibers, and more.) and organic particulate e.g. ceramic, fly ash, copy toner, and more.
The Air-O-Cell® collects both viable and non-viable sample specimens, providing a much broader overview of potential allergens and contaminants than conventional sampling techniques. After sampling is completed, the cassettes are sent to a laboratory, where the slides are removed and direct microscopic analysis can be immediately performed.
The collection media is compatible with a wide range of biological stains and refractive index oils, allowing direct quantitative analysis of organic and inorganic particulate. The Air-O-Cell® can be used with any standard off-the-shelf area sampling pump capable of drawing 15 LPM open flow. The small compact size makes Air-O-Cell® suitable for use in confined or restrictive area.
Spécifications
  • Désignation:
    Cassette