Microtube Rack, 4-Way, Electron Microscopy Sciences | Avantor
Microtube Rack, 4-Way, Electron Microscopy Sciences
4-Way Microtube Rack Assorted
Catalog # 102681-010
Fournisseur #:  61615-00
Microtube Rack, 4-Way, Electron Microscopy Sciences
Catalog # 102681-010
Microtube Rack, 4-Way, Electron Microscopy Sciences
Microtube Rack, 4-Way, Electron Microscopy Sciences
4-Way Microtube Rack Assorted
Price
$190.11
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Create as large a rack as you need by linking together multiple racks. Heavyduty polypropylene racks feature a unique system of tabs and slots, that facilitate easy connection and sturdy fit. Autoclavable. Assorted colors or natural. Measurements: 174 x 95 x 52 mm.
Spécifications
  • Désignation:
    4-Way Microtube Rack Assorted