100-Well Storage Racks, Electron Microscopy Sciences | Avantor
100-Well Storage Racks, Electron Microscopy Sciences
Natural
Catalog # 102680-992
Fournisseur #:  61601-N
100-Well Storage Racks, Electron Microscopy Sciences
Catalog # 102680-992
100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
Natural
Price
$144.71
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Polypropylene rack features durable three point hinges and clasp for secure lid closure.
  • Holds 1.5 to 2.0 ml microtubes and cryovials
  • Rack measures 5.625×5.875×2.25"
User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base and imprinted and raised coordinates on the box.
Spécifications
  • Color:
    Natural