100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
Blue
  102680-988
Supplier #:  61601-B
100-Well Storage Racks, Electron Microscopy Sciences
  102680-988
100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
Blue
153,04 $
Each Pack of 5
Polypropylene rack features durable three point hinges and clasp for secure lid closure.
  • Holds 1.5 to 2.0 ml microtubes and cryovials
  • Rack measures 5.625×5.875×2.25"
User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base and imprinted and raised coordinates on the box.
  • Color:
    Blue