Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific
250 mL
: J63974-AK
Phosphate, 0.5 M Buffer Solution, pH 7.2, Thermo Scientific
CAAAJ63974-AK
:
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
- Taille:250 mL
- Cat. No.:CAAAJ63974-AK
- MDL No.:MFCD00003527



