Sodium Borate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific
250 mL
Supplier: Thermo Scientific Chemicals
: J63637-AK
Sodium Borate, 0.5 M Buffer Solution, pH 9.0, Thermo Scientific
Catalog # CAAAJ63637-AK
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 1330-43-4
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00081185
Spécifications
Spécifications
- Taille:250 mL
- Cat. No.:CAAAJ63637-AK
- MDL No.:MFCD00081185



