MOBS, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
100 mL
Supplier: Thermo Scientific Chemicals
Fournisseur #: J63594-AE
MOBS, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Catalog # CAAAJ63594-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
CAS No.: 115724-21-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00674916
Spécifications
Spécifications
- Taille:100 mL
- Cat. No.:CAAAJ63594-AE
- MDL No.:MFCD00674916



