Stacking buffer (4X), Thermo Scientific
1 L
Supplier: Thermo Scientific Chemicals
: J63450-K2
Stacking buffer (4X), Thermo Scientific
Catalog # CAAAJ63450-K2
Supplier: Thermo Scientific Chemicals
CAS Number:
Jump to:
Product Details & Documents
Liquid
Physical Form: Liquid
Spécifications
Spécifications
- Taille:1 L



