Pectinase
250 mg
Supplier: Thermo Scientific Chemicals
Supplier #: J63408-MD
Jump to:
Product Details & Documents
Powder
CAS No.: 9032-75-1 Physical Form: Powder Appearance: Off-white to pale brown MDL No.: MFCD00131809
Specifications
Specifications
- Taille:250 mg
- Cat. No.:CAAAJ63408-MD
- Form:Powder
- Enzyme name:Pectinase
- CAS No.:9032-75-1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



